Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAAAACCAGAGCAATATACAAATA[C/T]TGTAATCACTTTTGGATTACAAAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606278 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FBXW7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FBXW7 - F-box and WD repeat domain containing 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001013415.1 | Intron | NP_001013433.1 | ||||
NM_001257069.1 | Intron | NP_001243998.1 | ||||
NM_018315.4 | Intron | NP_060785.2 | ||||
NM_033632.3 | Intron | NP_361014.1 | ||||
XM_011532083.1 | Intron | XP_011530385.1 | ||||
XM_011532084.1 | Intron | XP_011530386.1 | ||||
XM_011532085.1 | Intron | XP_011530387.1 | ||||
XM_011532086.1 | Intron | XP_011530388.1 | ||||
XM_011532087.1 | Intron | XP_011530389.1 | ||||
XM_011532088.1 | Intron | XP_011530390.1 | ||||
XM_017008362.1 | Intron | XP_016863851.1 |