Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGCGAATTATTCTACGGTTGTCAG[A/G]GTCATTCTGTTCCATATCTCCTCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
G3BP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
G3BP2 - G3BP stress granule assembly factor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012297.4 | 1115 | Missense Mutation | CCT,TCT | P,S 319 | NP_036429.2 | |
NM_203504.2 | 1115 | Missense Mutation | CCT,TCT | P,S 286 | NP_987100.1 | |
NM_203505.2 | 1115 | Missense Mutation | CCT,TCT | P,S 319 | NP_987101.1 | |
XM_005263382.2 | 1115 | Missense Mutation | CCT,TCT | P,S 319 | XP_005263439.1 | |
XM_005263383.3 | 1115 | Missense Mutation | CCT,TCT | P,S 319 | XP_005263440.1 | |
XM_011532441.2 | 1115 | Missense Mutation | CCT,TCT | P,S 319 | XP_011530743.1 | |
XM_017008876.1 | 1115 | Missense Mutation | CCT,TCT | P,S 319 | XP_016864365.1 | |
XM_017008877.1 | 1115 | Missense Mutation | CCT,TCT | P,S 319 | XP_016864366.1 | |
XM_017008878.1 | 1115 | Missense Mutation | CCT,TCT | P,S 286 | XP_016864367.1 | |
XM_017008879.1 | 1115 | Missense Mutation | CCT,TCT | P,S 286 | XP_016864368.1 |