Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTGCTGCCCGAGAAGCTCATAACC[A/G]AAGATGTCAGCCTCCCCTGTCCCGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611792 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZCCHC4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZCCHC4 - zinc finger CCHC-type containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318148.1 | 202 | Missense Mutation | CAA,CGA | Q,R 96 | NP_001305077.1 | |
NM_024936.2 | 202 | Missense Mutation | CAA,CGA | Q,R 96 | NP_079212.2 | |
XM_011513835.1 | 202 | Missense Mutation | CAA,CGA | Q,R 96 | XP_011512137.1 | |
XM_017008128.1 | 202 | Missense Mutation | CAA,CGA | Q,R 51 | XP_016863617.1 | |
XM_017008129.1 | 202 | Missense Mutation | CAA,CGA | Q,R 96 | XP_016863618.1 |