Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
GCTTCACTTCATACTCCCTTTCGAG[C/T]TTGATAGCTGAAAAAGAAAATACAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
C4orf36 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C4orf36 - chromosome 4 open reading frame 36 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_144645.3 | 428 | Silent Mutation | AAA,AAG | K,K 76 | NP_653246.1 | |
XM_011531610.2 | 428 | Silent Mutation | AAA,AAG | K,K 76 | XP_011529912.1 | |
XM_011531611.2 | 428 | Silent Mutation | AAA,AAG | K,K 76 | XP_011529913.1 | |
XM_011531612.2 | 428 | Silent Mutation | AAA,AAG | K,K 76 | XP_011529914.1 | |
XM_011531613.2 | 428 | Intron | XP_011529915.1 | |||
XM_011531615.2 | 428 | Silent Mutation | AAA,AAG | K,K 76 | XP_011529917.1 | |
XM_017007746.1 | 428 | Silent Mutation | AAA,AAG | K,K 76 | XP_016863235.1 |