Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAACAGGGAACCTGCTTCTTTCCA[A/G]TTGCACTTGGGTCTTTGCAAAAGTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601532 | ||||||||||||||||||||
Literature Links: |
CASP6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CASP6 - caspase 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001226.3 | 620 | Missense Mutation | NP_001217.2 | |||
NM_032992.2 | 620 | Missense Mutation | NP_116787.1 |
MCUB - mitochondrial calcium uniporter dominant negative beta subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017918.4 | 620 | Intron | NP_060388.2 | |||
XM_006714246.3 | 620 | Intron | XP_006714309.1 |