Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAATGTTCGGCTTAGCTGTTTGGT[A/G]GCAAATTGCAGTGCTCCATTTAGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606280 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DCAF16 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DCAF16 - DDB1 and CUL4 associated factor 16 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017741.3 | 1389 | Silent Mutation | GCC,GCT | A,A 148 | NP_060211.3 | |
XM_005248169.3 | 1389 | Silent Mutation | GCC,GCT | A,A 148 | XP_005248226.1 | |
XM_005248171.3 | 1389 | Silent Mutation | GCC,GCT | A,A 148 | XP_005248228.1 | |
XM_006713966.3 | 1389 | Silent Mutation | GCC,GCT | A,A 148 | XP_006714029.1 | |
XM_017008327.1 | 1389 | Silent Mutation | GCC,GCT | A,A 148 | XP_016863816.1 | |
XM_017008328.1 | 1389 | Silent Mutation | GCC,GCT | A,A 148 | XP_016863817.1 |
NCAPG - non-SMC condensin I complex subunit G | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |