Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATCCCCACTATCCTGGTGACCTCT[A/G]TGACCTCTGAGACCCCAGCAACAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604552 MIM: 602512 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HGFAC PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HGFAC - HGF activator | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001297439.1 | 256 | Missense Mutation | ATG,GTG | M,V 63 | NP_001284368.1 | |
NM_001528.3 | 256 | Missense Mutation | ATG,GTG | M,V 63 | NP_001519.1 |
RGS12 - regulator of G-protein signaling 12 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |