Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAACTTGAGGCTCCTTTCACTCAGG[A/G]TGACACCCTTGGACTGGAAAACTCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605303 MIM: 615975 | ||||||||||||||||||||
Literature Links: |
TACC3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TACC3 - transforming acidic coiled-coil containing protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006342.2 | 407 | Missense Mutation | GAT,GGT | D,G 85 | NP_006333.1 | |
XM_005247929.2 | 407 | Missense Mutation | GAT,GGT | D,G 85 | XP_005247986.1 | |
XM_005247930.2 | 407 | Missense Mutation | GAT,GGT | D,G 85 | XP_005247987.1 | |
XM_011513386.2 | 407 | Missense Mutation | GAT,GGT | D,G 85 | XP_011511688.1 | |
XM_017007653.1 | 407 | Missense Mutation | GAT,GGT | D,G 85 | XP_016863142.1 |
TMEM129 - transmembrane protein 129 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |