Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCATCTTCCTGTTGAGCTGTACTT[A/C]GGTTTATGGCCTTTTCCATTCCGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 102680 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ADD1 - adducin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001119.4 | Intron | NP_001110.2 | ||||
NM_001286645.1 | Intron | NP_001273574.1 | ||||
NM_014189.3 | Intron | NP_054908.2 | ||||
NM_014190.3 | Intron | NP_054909.2 | ||||
NM_176801.2 | Intron | NP_789771.1 | ||||
XM_005247933.2 | Intron | XP_005247990.1 | ||||
XM_005247934.2 | Intron | XP_005247991.1 | ||||
XM_005247935.2 | Intron | XP_005247992.1 | ||||
XM_005247936.2 | Intron | XP_005247993.1 | ||||
XM_005247937.2 | Intron | XP_005247994.1 | ||||
XM_017007703.1 | Intron | XP_016863192.1 | ||||
XM_017007704.1 | Intron | XP_016863193.1 | ||||
XM_017007705.1 | Intron | XP_016863194.1 | ||||
XM_017007706.1 | Intron | XP_016863195.1 | ||||
XM_017007707.1 | Intron | XP_016863196.1 | ||||
XM_017007708.1 | Intron | XP_016863197.1 | ||||
XM_017007709.1 | Intron | XP_016863198.1 | ||||
XM_017007710.1 | Intron | XP_016863199.1 |