Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTCACCTGTTTTTCTCGGAAGGAA[C/T]GCTTATTTTTTGACCGGACATTCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
SDAD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SDAD1 - SDA1 domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001288983.1 | 2174 | Intron | NP_001275912.1 | |||
NM_001288984.1 | 2174 | Intron | NP_001275913.1 | |||
NM_018115.3 | 2174 | Intron | NP_060585.2 | |||
XM_005263102.4 | 2174 | Missense Mutation | CAT,CGT | H,R 641 | XP_005263159.1 | |
XM_005263105.4 | 2174 | Intron | XP_005263162.1 | |||
XM_017008354.1 | 2174 | Intron | XP_016863843.1 | |||
XM_017008355.1 | 2174 | Intron | XP_016863844.1 |