Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTTCCATTATTTCAGGCTGAGGGA[A/C]AGGCAGCACCACAGCAGGCTGAGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 115460 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CSN2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CSN2 - casein beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001302770.1 | 293 | Missense Mutation | GTC,TTC | V,F 92 | NP_001289699.1 | |
NM_001891.3 | 293 | Missense Mutation | GTC,TTC | V,F 93 | NP_001882.1 | |
XM_017007760.1 | 293 | Missense Mutation | GTC,TTC | V,F 93 | XP_016863249.1 |