Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCACCGGGGAGGACATAGCTGTTGC[A/G]CAAAATCCAGCGATAAGAGCCGACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610196 MIM: 113730 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ELMOD2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ELMOD2 - ELMO domain containing 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UCP1 - uncoupling protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021833.4 | 751 | Silent Mutation | TGC,TGT | C,C 225 | NP_068605.1 | |
XM_005263206.3 | 751 | Silent Mutation | TGC,TGT | C,C 224 | XP_005263263.1 | |
XM_011532228.2 | 751 | Silent Mutation | TGC,TGT | C,C 225 | XP_011530530.1 |