Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGATATGACATTCGCTTGCGGCCG[A/G]ACTTCGGAGGTAACGCTTCATCTTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 137190 | ||||||||||||||||||||
Literature Links: |
GABRB1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GABRB1 - gamma-aminobutyric acid type A receptor beta1 subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000812.3 | 262 | Missense Mutation | AAC,GAC | N,D 55 | NP_000803.2 | |
XM_017007985.1 | 262 | Intron | XP_016863474.1 | |||
XM_017007986.1 | 262 | Missense Mutation | AAC,GAC | N,D 55 | XP_016863475.1 |