Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCACTTGCCAAGGAAGTGCCACACT[C/T]GCTCTGCATTTAAAATGGGGAAAGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609988 | ||||||||||||||||||||
Literature Links: |
PPA2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PPA2 - pyrophosphatase (inorganic) 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006903.4 | 913 | Missense Mutation | CAA,CGA | Q,R 298 | NP_008834.3 | |
NM_176866.2 | 913 | Missense Mutation | CAA,CGA | Q,R 225 | NP_789842.2 | |
NM_176867.3 | 913 | Missense Mutation | CAA,CGA | Q,R 161 | NP_789843.2 | |
NM_176869.2 | 913 | Missense Mutation | CAA,CGA | Q,R 327 | NP_789845.1 |