Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCTCAATCAGCTAGGCTCAACACA[T/C]AGAAGCTTAGAACTAGAAATGACAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607494 | ||||||||||||||||||||
Literature Links: |
INPP4B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
INPP4B - inositol polyphosphate-4-phosphatase type II B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001101669.1 | 3604 | UTR 3 | NP_001095139.1 | |||
NM_003866.2 | 3604 | UTR 3 | NP_003857.2 | |||
XM_005263323.3 | 3604 | Intron | XP_005263380.1 | |||
XM_005263325.3 | 3604 | Intron | XP_005263382.1 | |||
XM_011532391.2 | 3604 | Intron | XP_011530693.1 | |||
XM_011532392.2 | 3604 | Intron | XP_011530694.1 | |||
XM_017008794.1 | 3604 | UTR 3 | XP_016864283.1 | |||
XM_017008795.1 | 3604 | Intron | XP_016864284.1 | |||
XM_017008796.1 | 3604 | Intron | XP_016864285.1 | |||
XM_017008797.1 | 3604 | Intron | XP_016864286.1 | |||
XM_017008798.1 | 3604 | Intron | XP_016864287.1 |