Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGAAGAGTGTAGCATTTTTGGAAG[A/G]AGAAATTCCCATTTTAAGTCAACCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616523 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MFAP3L PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MFAP3L - microfibrillar associated protein 3 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001009554.3 | 4098 | UTR 3 | NP_001009554.1 | |||
NM_001301647.1 | 4098 | UTR 3 | NP_001288576.1 | |||
NM_001301648.1 | 4098 | UTR 3 | NP_001288577.1 | |||
NM_021647.7 | 4098 | UTR 3 | NP_067679.6 | |||
XM_005263366.3 | 4098 | UTR 3 | XP_005263423.1 | |||
XM_005263368.3 | 4098 | UTR 3 | XP_005263425.1 | |||
XM_017008864.1 | 4098 | UTR 3 | XP_016864353.1 | |||
XM_017008865.1 | 4098 | UTR 3 | XP_016864354.1 | |||
XM_017008866.1 | 4098 | UTR 3 | XP_016864355.1 | |||
XM_017008867.1 | 4098 | UTR 3 | XP_016864356.1 | |||
XM_017008868.1 | 4098 | UTR 3 | XP_016864357.1 | |||
XM_017008869.1 | 4098 | UTR 3 | XP_016864358.1 | |||
XM_017008870.1 | 4098 | UTR 3 | XP_016864359.1 | |||
XM_017008871.1 | 4098 | UTR 3 | XP_016864360.1 |