Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTCTGGGCAGCCACCCGGTCCCC[C/T]TCAGGCCCCGCCGGGCCTTCCGGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 170250 MIM: 610311 | ||||||||||||||||||||
Literature Links: |
LAP3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LAP3 - leucine aminopeptidase 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MED28 - mediator complex subunit 28 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_025205.4 | 77 | Missense Mutation | CCT,CTT | P,L 17 | NP_079481.2 |