Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAAGGCAGATTGCATGGGCCCTTC[A/G]AGATCTTGCTGTGGTAGATGCCAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607879 | ||||||||||||||||||||
Literature Links: |
EXOC1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
EXOC1 - exocyst complex component 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001024924.1 | 376 | Missense Mutation | CAA,CGA | Q,R 73 | NP_001020095.1 | |
NM_018261.3 | 376 | Missense Mutation | CAA,CGA | Q,R 73 | NP_060731.2 | |
NM_178237.2 | 376 | Missense Mutation | CAA,CGA | Q,R 73 | NP_839955.1 | |
XM_005265747.2 | 376 | Missense Mutation | CAA,CGA | Q,R 73 | XP_005265804.1 | |
XM_005265748.1 | 376 | Missense Mutation | CAA,CGA | Q,R 73 | XP_005265805.1 | |
XM_005265750.2 | 376 | Missense Mutation | CAA,CGA | Q,R 73 | XP_005265807.1 | |
XM_017008405.1 | 376 | Missense Mutation | CAA,CGA | Q,R 73 | XP_016863894.1 | |
XM_017008406.1 | 376 | Missense Mutation | CAA,CGA | Q,R 73 | XP_016863895.1 | |
XM_017008407.1 | 376 | Missense Mutation | CAA,CGA | Q,R 73 | XP_016863896.1 | |
XM_017008408.1 | 376 | Intron | XP_016863897.1 | |||
XM_017008409.1 | 376 | Intron | XP_016863898.1 |