Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGAGGTGGGAGGAAAGGGGTCATT[A/G]CCACTCCTGGATTCATGTTCTACCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 153245 | ||||||||||||||||||||
Literature Links: |
LEF1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LEF1 - lymphoid enhancer binding factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130713.2 | Intron | NP_001124185.1 | ||||
NM_001130714.2 | Intron | NP_001124186.1 | ||||
NM_001166119.1 | Intron | NP_001159591.1 | ||||
NM_016269.4 | Intron | NP_057353.1 | ||||
XM_005263046.3 | Intron | XP_005263103.1 | ||||
XM_005263047.1 | Intron | XP_005263104.1 | ||||
XM_005263048.1 | Intron | XP_005263105.1 | ||||
XM_006714233.1 | Intron | XP_006714296.1 |