Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTAAAGAGAGCAAGATGTCGTTTGA[A/G]CTTTTTCTTCTCCTCTGTCTGTTCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606925 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
QRFPR PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
QRFPR - pyroglutamylated RFamide peptide receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198179.2 | 1607 | Missense Mutation | CTC,TTC | L,F 410 | NP_937822.2 | |
XM_017008693.1 | 1607 | Intron | XP_016864182.1 |