Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCTCTACGCTGCGGGGATGCAGCC[C/T]GCACCCTGGGGCCCCGGGTGAGCGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611135 MIM: 607031 MIM: 603686 | ||||||||||||||||||||
Literature Links: |
KLB PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KLB - klotho beta | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LIAS - lipoic acid synthetase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278590.1 | 122 | Missense Mutation | CGC,TGC | R,C 10 | NP_001265519.1 | |
NM_001278591.1 | 122 | Missense Mutation | CGC,TGC | R,C 10 | NP_001265520.1 | |
NM_001278592.1 | 122 | Missense Mutation | CGC,TGC | R,C 10 | NP_001265521.1 | |
NM_006859.3 | 122 | Missense Mutation | CGC,TGC | R,C 10 | NP_006850.2 | |
NM_194451.2 | 122 | Missense Mutation | CGC,TGC | R,C 10 | NP_919433.1 | |
XM_006713990.3 | 122 | Missense Mutation | CGC,TGC | R,C 10 | XP_006714053.1 | |
XM_017007665.1 | 122 | Missense Mutation | CGC,TGC | R,C 10 | XP_016863154.1 | |
XM_017007666.1 | 122 | Missense Mutation | CGC,TGC | R,C 10 | XP_016863155.1 | |
XM_017007667.1 | 122 | Missense Mutation | CGC,TGC | R,C 10 | XP_016863156.1 |
RPL9 - ribosomal protein L9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000661.4 | 122 | Intron | NP_000652.2 | |||
NM_001024921.3 | 122 | Intron | NP_001020092.1 |