Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACCAACAAGTTCATCAAAGAGCTC[A/G]TTAAGGACGGGAAGAACCTCATCGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609746 | ||||||||||||||||||||
Literature Links: |
ARHGAP10 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ARHGAP10 - Rho GTPase activating protein 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024605.3 | 354 | Missense Mutation | ATT,GTT | I,V 40 | NP_078881.3 | |
XM_005263215.3 | 354 | Missense Mutation | ATT,GTT | I,V 40 | XP_005263272.1 | |
XM_017008602.1 | 354 | UTR 5 | XP_016864091.1 |