Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAGATGAACAGGGAACTAGAATTG[C/T]TGGAAATTCTAAGTTACTTGTAATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609825 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
COQ2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
COQ2 - coenzyme Q2, polyprenyltransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015697.7 | Intron | NP_056512.5 | ||||
XM_011531855.2 | Intron | XP_011530157.1 | ||||
XM_011531857.2 | Intron | XP_011530159.1 | ||||
XM_011531859.2 | Intron | XP_011530161.1 | ||||
XM_011531860.2 | Intron | XP_011530162.1 | ||||
XM_011531862.2 | Intron | XP_011530164.1 | ||||
XM_011531863.2 | Intron | XP_011530165.1 | ||||
XM_011531866.2 | Intron | XP_011530168.1 | ||||
XM_011531867.2 | Intron | XP_011530169.1 | ||||
XM_017008031.1 | Intron | XP_016863520.1 | ||||
XM_017008032.1 | Intron | XP_016863521.1 |