Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATGGATTAAAAAAAAAAGCTCACC[G/T]TTGGTATACAGATAATGACAAAATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613367 MIM: 612537 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LIN54 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LIN54 - lin-54 DREAM MuvB core complex component | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001115007.2 | 5381 | UTR 3 | NP_001108479.1 | |||
NM_001115008.2 | 5381 | UTR 3 | NP_001108480.1 | |||
NM_001288996.1 | 5381 | UTR 3 | NP_001275925.1 | |||
NM_001288997.1 | 5381 | UTR 3 | NP_001275926.1 | |||
NM_194282.3 | 5381 | UTR 3 | NP_919258.2 | |||
XM_005262750.4 | 5381 | Intron | XP_005262807.1 | |||
XM_006714081.3 | 5381 | Intron | XP_006714144.1 | |||
XM_017007728.1 | 5381 | Intron | XP_016863217.1 | |||
XM_017007729.1 | 5381 | Intron | XP_016863218.1 | |||
XM_017007730.1 | 5381 | Intron | XP_016863219.1 |
THAP9 - THAP domain containing 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |