Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTGCTCCATCAGCAGGGTCCATTT[C/T]GAATGATTTTAATATTATGACCACC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605753 | ||||||||||||||||||||
Literature Links: |
SPINK2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SPINK2 - serine peptidase inhibitor, Kazal type 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271718.1 | 560 | Missense Mutation | CAA,CGA | Q,R 130 | NP_001258647.1 | |
NM_001271719.1 | 560 | UTR 3 | NP_001258648.1 | |||
NM_001271720.1 | 560 | Missense Mutation | CAA,CGA | Q,R 115 | NP_001258649.1 | |
NM_001271721.1 | 560 | UTR 3 | NP_001258650.1 | |||
NM_001271722.1 | 560 | Missense Mutation | AAA,GAA | K,E 79 | NP_001258651.1 | |
NM_021114.3 | 560 | Missense Mutation | CAA,CGA | Q,R 80 | NP_066937.1 | |
XM_011534405.1 | 560 | Missense Mutation | CAA,CGA | Q,R 182 | XP_011532707.1 | |
XM_011534406.2 | 560 | Intron | XP_011532708.1 |