Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTACAGCCGTCACATTGTCCTTGC[C/T]TGCCCCTGAGCTCTCGTCCTGCAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
LRRC66 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LRRC66 - leucine rich repeat containing 66 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001024611.2 | 3961 | Missense Mutation | AGC,GGC | S,G 743 | NP_001019782.1 | |
XM_011534383.2 | 3961 | Missense Mutation | AGC,GGC | S,G 795 | XP_011532685.1 | |
XM_017008162.1 | 3961 | Missense Mutation | AGC,GGC | S,G 507 | XP_016863651.1 |