Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTGGCTTCAGGAAGACAAATCTTA[C/T]GAATGTACTCTGTAAAAGAAACTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
TMPRSS11B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TMPRSS11B - transmembrane protease, serine 11B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_182502.3 | 832 | Missense Mutation | CAT,CGT | H,R 287 | NP_872308.2 | |
XM_011531608.2 | 832 | Missense Mutation | CAT,CGT | H,R 287 | XP_011529910.1 | |
XM_011531609.1 | 832 | Silent Mutation | TCA,TCG | S,S 232 | XP_011529911.1 |