Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGACATCATATTCTCCAATTCCACA[A/G]TACCATTCCAGGTAGACTATGCAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
TMEM128 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TMEM128 - transmembrane protein 128 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001297551.1 | 358 | Silent Mutation | TAC,TAT | Y,Y 106 | NP_001284480.1 | |
NM_001297552.1 | 358 | Silent Mutation | TAC,TAT | Y,Y 106 | NP_001284481.1 | |
NM_032927.3 | 358 | Silent Mutation | TAC,TAT | Y,Y 82 | NP_116316.1 | |
XM_005248034.2 | 358 | Silent Mutation | TAC,TAT | Y,Y 106 | XP_005248091.1 |