Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATTGGCTGAACAGAACTGTATTCA[A/G]TAATTCATATTCTATAACTTCATTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613009 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SEPSECS PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SEPSECS - Sep (O-phosphoserine) tRNA:Sec (selenocysteine) tRNA synthase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016955.3 | 4533 | UTR 3 | NP_058651.3 | |||
XM_011513846.2 | 4533 | UTR 3 | XP_011512148.1 | |||
XM_011513847.2 | 4533 | UTR 3 | XP_011512149.1 | |||
XM_011513848.1 | 4533 | UTR 3 | XP_011512150.1 | |||
XM_017008277.1 | 4533 | UTR 3 | XP_016863766.1 | |||
XM_017008278.1 | 4533 | UTR 3 | XP_016863767.1 |