Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGGAAGCTGAAGGCTGAGAGTCTC[C/T]CTTCTCACTTAATCCATGCTTTATT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
OCIAD2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
OCIAD2 - OCIA domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001014446.2 | 577 | Missense Mutation | NP_001014446.1 | |||
NM_001286773.1 | 577 | UTR 3 | NP_001273702.1 | |||
NM_001286774.1 | 577 | Missense Mutation | NP_001273703.1 | |||
NM_152398.3 | 577 | UTR 3 | NP_689611.1 |