Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACCACAGCTGGCTTCCTTAACGCA[C/T]CTTTTTTACTAGTGGTTGAAACTGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612303 | ||||||||||||||||||||
Literature Links: |
ADGRA3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ADGRA3 - adhesion G protein-coupled receptor A3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145290.3 | 4061 | Missense Mutation | GAT,GGT | D,G 1264 | NP_660333.2 | |
XM_005248137.4 | 4061 | Intron | XP_005248194.1 | |||
XM_011513811.2 | 4061 | Missense Mutation | GAT,GGT | D,G 1038 | XP_011512113.1 | |
XM_017007834.1 | 4061 | Missense Mutation | GAT,GGT | D,G 1067 | XP_016863323.1 |