Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTCACCATCAGTTTTTAGAAAGTA[C/T]GCATGGCTCTCCTTCTGTTGATATC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607515 | ||||||||||||||||||||
Literature Links: |
PLAC8 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PLAC8 - placenta specific 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130715.1 | 499 | Missense Mutation | CAT,CGT | H,R 113 | NP_001124187.1 | |
NM_001130716.1 | 499 | Missense Mutation | CAT,CGT | H,R 113 | NP_001124188.1 | |
NM_016619.2 | 499 | Missense Mutation | CAT,CGT | H,R 113 | NP_057703.1 |