Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATAGAATGGAATGGGTTCCAGTAAG[A/G]TAAGAATTATATGATCAGAATTTTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606270 | ||||||||||||||||||||
Literature Links: |
TLR10 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TLR10 - toll like receptor 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001017388.2 | 2315 | Missense Mutation | ACC,ATC | T,I 728 | NP_001017388.1 | |
NM_001195106.1 | 2315 | Missense Mutation | ACC,ATC | T,I 728 | NP_001182035.1 | |
NM_001195107.1 | 2315 | Missense Mutation | ACC,ATC | T,I 728 | NP_001182036.1 | |
NM_001195108.1 | 2315 | Missense Mutation | ACC,ATC | T,I 714 | NP_001182037.1 | |
NM_030956.3 | 2315 | Missense Mutation | ACC,ATC | T,I 728 | NP_112218.2 | |
XM_011513760.2 | 2315 | Missense Mutation | ACC,ATC | T,I 714 | XP_011512062.1 | |
XM_011513761.2 | 2315 | Missense Mutation | ACC,ATC | T,I 728 | XP_011512063.1 | |
XM_011513762.2 | 2315 | Missense Mutation | ACC,ATC | T,I 728 | XP_011512064.1 |