Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGAGGAAATAAATATCTTTGATTA[A/C]GAGACTGAAGAATGGTATCACAATC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614532 | ||||||||||||||||||||
Literature Links: |
RASGEF1B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RASGEF1B - RasGEF domain family member 1B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300735.1 | 1294 | Missense Mutation | GTA,TTA | V,L 376 | NP_001287664.1 | |
NM_001300736.1 | 1294 | Missense Mutation | GTA,TTA | V,L 335 | NP_001287665.1 | |
NM_152545.2 | 1294 | Missense Mutation | GTA,TTA | V,L 377 | NP_689758.1 | |
XM_017007812.1 | 1294 | Missense Mutation | GTA,TTA | V,L 336 | XP_016863301.1 | |
XM_017007813.1 | 1294 | Missense Mutation | GTA,TTA | V,L 377 | XP_016863302.1 |