Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAAAGTGTTTGATATTCCAAGTGA[C/T]GAATGTATTAAGGTAAAATACGTTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
GUF1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GUF1 - GUF1 homolog, GTPase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021927.2 | 718 | Silent Mutation | GAC,GAT | D,D 219 | NP_068746.2 | |
XM_005248122.3 | 718 | UTR 5 | XP_005248179.1 | |||
XM_011513732.2 | 718 | Silent Mutation | GAC,GAT | D,D 219 | XP_011512034.1 |