Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCACTATCTTTGGAGGAAGTCCTAA[C/T]CTTTCCTTGTATACCCTCCTAGAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 133440 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EIF4E PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EIF4E - eukaryotic translation initiation factor 4E | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130678.1 | 2081 | Silent Mutation | AGA,AGG | R,R 206 | NP_001124150.1 | |
NM_001130679.1 | 2081 | Silent Mutation | AGA,AGG | R,R 217 | NP_001124151.1 | |
NM_001968.3 | 2081 | Silent Mutation | AGA,AGG | R,R 186 | NP_001959.1 | |
XM_006714126.3 | 2081 | Silent Mutation | AGA,AGG | R,R 214 | XP_006714189.1 |