Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTGTTCACGGTGGCTTTAATTTTC[C/T]TGCCTAACGTTGGTCTCTGGTCTCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615138 | ||||||||||||||||||||
Literature Links: |
GALNTL6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GALNTL6 - polypeptide N-acetylgalactosaminyltransferase-like 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001034845.2 | 800 | Silent Mutation | CTG,TTG | L,L 22 | NP_001030017.2 | |
XM_011531993.2 | 800 | Intron | XP_011530295.1 | |||
XM_011531994.1 | 800 | Intron | XP_011530296.1 | |||
XM_011531995.1 | 800 | Intron | XP_011530297.1 | |||
XM_011531996.1 | 800 | Intron | XP_011530298.1 | |||
XM_011531997.1 | 800 | Intron | XP_011530299.1 | |||
XM_017008243.1 | 800 | Silent Mutation | CTG,TTG | L,L 22 | XP_016863732.1 | |
XM_017008244.1 | 800 | Intron | XP_016863733.1 |
LOC441052 - uncharacterized LOC441052 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |