Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCGAATGAGGGGTGCTCTTTCATC[C/T]GCTGTTCCCTGAAACACAGAAGAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602257 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SCARB2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SCARB2 - scavenger receptor class B member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204255.1 | 1334 | Silent Mutation | GCA,GCG | A,A 326 | NP_001191184.1 | |
NM_005506.3 | 1334 | Silent Mutation | GCA,GCG | A,A 469 | NP_005497.1 |