Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCCCCAGTGGCATCACTCCCCAT[C/T]TGGGGAGTTCAGGGTCACCCTGCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608991 MIM: 601733 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAML3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MAML3 - mastermind like transcriptional coactivator 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018717.4 | 6038 | UTR 3 | NP_061187.2 |
MGST2 - microsomal glutathione S-transferase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204366.1 | 6038 | Intron | NP_001191295.1 | |||
NM_001204367.1 | 6038 | Intron | NP_001191296.1 | |||
NM_001204368.1 | 6038 | Intron | NP_001191297.1 | |||
NM_002413.4 | 6038 | Intron | NP_002404.1 | |||
XM_006714221.2 | 6038 | Intron | XP_006714284.1 | |||
XM_011531972.2 | 6038 | Intron | XP_011530274.1 | |||
XM_017008211.1 | 6038 | Intron | XP_016863700.1 | |||
XM_017008212.1 | 6038 | Intron | XP_016863701.1 | |||
XM_017008213.1 | 6038 | Intron | XP_016863702.1 | |||
XM_017008214.1 | 6038 | Intron | XP_016863703.1 |