Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTCATCGATTGGCCTATGTCGTTA[C/T]GGTGGGAGGATTGACTGCTGCTGGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610306 | ||||||||||||||||||||
Literature Links: |
NPNT PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NPNT - nephronectin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001033047.2 | 327 | Silent Mutation | TAC,TAT | Y,Y 38 | NP_001028219.1 | |
NM_001184690.1 | 327 | Silent Mutation | TAC,TAT | Y,Y 38 | NP_001171619.1 | |
NM_001184691.1 | 327 | Silent Mutation | TAC,TAT | Y,Y 38 | NP_001171620.1 | |
NM_001184692.1 | 327 | Silent Mutation | TAC,TAT | Y,Y 38 | NP_001171621.1 | |
NM_001184693.1 | 327 | Silent Mutation | TAC,TAT | Y,Y 38 | NP_001171622.1 | |
XM_005262888.4 | 327 | Silent Mutation | TAC,TAT | Y,Y 38 | XP_005262945.1 | |
XM_005262890.1 | 327 | Intron | XP_005262947.1 | |||
XM_011531820.2 | 327 | Silent Mutation | TAC,TAT | Y,Y 39 | XP_011530122.1 | |
XM_011531822.2 | 327 | Silent Mutation | TAC,TAT | Y,Y 39 | XP_011530124.1 | |
XM_011531823.2 | 327 | Silent Mutation | TAC,TAT | Y,Y 39 | XP_011530125.1 | |
XM_011531824.2 | 327 | Silent Mutation | TAC,TAT | Y,Y 39 | XP_011530126.1 | |
XM_011531825.2 | 327 | Silent Mutation | TAC,TAT | Y,Y 39 | XP_011530127.1 | |
XM_017007984.1 | 327 | Silent Mutation | TAC,TAT | Y,Y 39 | XP_016863473.1 |