Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTGGCGGGCTCGTGCTGTCTGCAC[A/G]GCATCGTGCACACTGAGGAACAGCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 252800 MIM: 610130 | ||||||||||||||||||||
Literature Links: |
IDUA PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IDUA - iduronidase, alpha-L- | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000203.4 | 2216 | Intron | NP_000194.2 | |||
XM_011513461.2 | 2216 | Intron | XP_011511763.1 | |||
XM_017008163.1 | 2216 | Intron | XP_016863652.1 |
SLC26A1 - solute carrier family 26 member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022042.3 | 2216 | Silent Mutation | GCC,GCT | A,A 683 | NP_071325.2 | |
NM_134425.2 | 2216 | Intron | NP_602297.1 | |||
NM_213613.3 | 2216 | Silent Mutation | GCC,GCT | A,A 683 | NP_998778.1 |