Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCATGATTAACTGCCAAAACTTTT[C/T]GACTATTCATCTTCACAAAATTTCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616013 | ||||||||||||||||||||
Literature Links: |
C4orf17 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C4orf17 - chromosome 4 open reading frame 17 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TRMT10A - tRNA methyltransferase 10A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001134665.2 | 875 | Missense Mutation | NP_001128137.1 | |||
NM_001134666.2 | 875 | Missense Mutation | NP_001128138.1 | |||
NM_152292.4 | 875 | Missense Mutation | NP_689505.1 | |||
XM_005263352.3 | 875 | Missense Mutation | XP_005263409.1 | |||
XM_006714417.2 | 875 | Missense Mutation | XP_006714480.1 |