Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCATTTCTTATCACAAAAAAACT[A/G]TATGAGAAAGCTGGCAAGCCCAGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604724 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HPSE PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HPSE - heparanase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098540.2 | 1502 | Silent Mutation | TAC,TAT | Y,Y 529 | NP_001092010.1 | |
NM_001166498.2 | 1502 | Silent Mutation | TAC,TAT | Y,Y 455 | NP_001159970.1 | |
NM_001199830.1 | 1502 | Silent Mutation | TAC,TAT | Y,Y 471 | NP_001186759.1 | |
NM_006665.5 | 1502 | Silent Mutation | TAC,TAT | Y,Y 529 | NP_006656.2 |