Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGAAGCAATATCCTTTCCACCATA[C/T]TGCTAGACGTAGCTACATCTATAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615050 | ||||||||||||||||||||
Literature Links: |
ASB5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ASB5 - ankyrin repeat and SOCS box containing 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_080874.3 | 1635 | Missense Mutation | AAT,AGT | N,S 277 | NP_543150.1 | |
XM_005262759.1 | 1635 | Missense Mutation | AAT,AGT | N,S 277 | XP_005262816.1 | |
XM_011531616.2 | 1635 | Missense Mutation | AAT,AGT | N,S 232 | XP_011529918.1 | |
XM_011531617.2 | 1635 | Missense Mutation | AAT,AGT | N,S 224 | XP_011529919.1 |