Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATACAACCTAGATTCCTTCAGTAA[A/G]TATCTGTTAAGGAGGTCACATGAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609489 | ||||||||||||||||||||
Literature Links: |
MANBA PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MANBA - mannosidase beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005908.3 | 1868 | Missense Mutation | ACT,ATT | T,I 878 | NP_005899.3 | |
XM_011531966.2 | 1868 | Missense Mutation | ACT,ATT | T,I 463 | XP_011530268.1 | |
XM_017008203.1 | 1868 | Missense Mutation | ACT,ATT | T,I 757 | XP_016863692.1 | |
XM_017008204.1 | 1868 | Missense Mutation | ACT,ATT | T,I 662 | XP_016863693.1 | |
XM_017008205.1 | 1868 | Missense Mutation | ACT,ATT | T,I 476 | XP_016863694.1 |