Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTTTGTCTCTTCAGGAAATCATTC[C/T]GTGCCTGTGACCAGAGACCATACCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611789 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BDH2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BDH2 - 3-hydroxybutyrate dehydrogenase, type 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020139.3 | Intron | NP_064524.3 | ||||
XM_005263140.3 | Intron | XP_005263197.1 | ||||
XM_006714274.3 | Intron | XP_006714337.1 | ||||
XM_011532127.2 | Intron | XP_011530429.1 | ||||
XM_011532128.2 | Intron | XP_011530430.1 | ||||
XM_017008461.1 | Intron | XP_016863950.1 | ||||
XM_017008462.1 | Intron | XP_016863951.1 |
SLC9B2 - solute carrier family 9 member B2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |