Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGCTTTTTTATTTCTATGACTTAC[C/G]TGCAACTTTGGGCAATGATCCAAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
OCIAD2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
OCIAD2 - OCIA domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001014446.2 | 521 | Missense Mutation | CTT,GTT | L,V 89 | NP_001014446.1 | |
NM_001286773.1 | 521 | Missense Mutation | CGC,GGC | R,G 89 | NP_001273702.1 | |
NM_001286774.1 | 521 | Missense Mutation | CTT,GTT | L,V 27 | NP_001273703.1 | |
NM_152398.3 | 521 | Missense Mutation | CGC,GGC | R,G 89 | NP_689611.1 |