Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCAGCCCAACCCCGATAAGTACCT[C/T]GAAGGGGCCGCAGGTCAGCAGCCCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604217 | ||||||||||||||||||||
Literature Links: |
SLC34A2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SLC34A2 - solute carrier family 34 member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001177998.1 | 158 | Silent Mutation | CTC,CTT | L,L 18 | NP_001171469.1 | |
NM_001177999.1 | 158 | Silent Mutation | CTC,CTT | L,L 18 | NP_001171470.1 | |
NM_006424.2 | 158 | Silent Mutation | CTC,CTT | L,L 18 | NP_006415.2 |