Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTAGTAACTTTGCTTGGGACACA[C/T]TGACATTCATTGCAATTGTGGAGAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608452 | ||||||||||||||||||||
Literature Links: |
PDGFC PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PDGFC - platelet derived growth factor C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016205.2 | 1005 | Silent Mutation | CAA,CAG | Q,Q 295 | NP_057289.1 | |
XM_011532124.2 | 1005 | Silent Mutation | CAA,CAG | Q,Q 284 | XP_011530426.1 | |
XM_017008455.1 | 1005 | Silent Mutation | CAA,CAG | Q,Q 260 | XP_016863944.1 | |
XM_017008456.1 | 1005 | Intron | XP_016863945.1 |