Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATTGTTGGTGGCCAGAACTCAAAT[C/T]ACATACAGTCAACCACATTATTGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611967 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KLHL8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KLHL8 - kelch like family member 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001292003.1 | 2204 | Silent Mutation | TAA,TGA | *,* 621 | NP_001278932.1 | |
NM_001292006.1 | 2204 | Silent Mutation | TAA,TGA | *,* 545 | NP_001278935.1 | |
NM_001292007.1 | 2204 | Silent Mutation | TAA,TGA | *,* 438 | NP_001278936.1 | |
NM_020803.4 | 2204 | Silent Mutation | TAA,TGA | *,* 621 | NP_065854.3 |